missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Zonisamide

Product Code. 18722624
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18722624 50 mg 50mg
18702614 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18722624 Supplier R&D Systems™ Supplier No. 2625/50
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Blocks voltage-sensitive Na+ and Ca2+ channels (T-type)

Antiepileptic that possesses a broad spectrum anticonvulsant and mechanistic profile. Blocks voltage-sensitive Na+ and T-type Ca2+ channels, stimulates BKCa channels, modulates GABA, glutamate and monoamine neurotransmission, inhibits lipid peroxidation and scavenges hydroxyl and nitric oxide free radicals. Displays neuroprotective and antiParkinsonian activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68291-97-4
Molecular Formula C8H8N2O3S

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 212.22
Purity >99%
Molecular Weight (g/mol) 212.22
Chemical Name or Material Zonisamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.