missing translation for 'onlineSavingsMsg'
Learn More

Zolpidem Phenyl-4-carboxylic Acid, TRC

Product Code. 30010047
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010047 1 mg 1mg
30005755 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010047 Supplier Toronto Research Chemicals Supplier No. TRCZ6500151MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109461-65-6
Molecular Formula C19 H19 N3 O3
Molecular Weight (g/mol) 337.37
Synonym Zolpidem Phenyl-4-carboxylic Acid, Benzoic acid, 4-[3-[2-(dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]-, 4-[3-[2-(Dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic acid, Imidazo[1,2-a]pyridine, benzoic acid deriv., SL 84.0589, Zolpidem carboxylic acid
IUPAC Name 4-[3-[2-(dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic acid
SMILES CN(C)C(=O)Cc1c(nc2ccc(C)cn12)c3ccc(cc3)C(=O)O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C19H19N3O3/c1-12-4-9-16-20-18(13-5-7-14(8-6-13)19(24)25)15(22(16)11-12)10-17(23)21(2)3/h4-9,11H,10H2,1-3H3,(H,24,25)
Formula Weight 337.1426
Percent Purity >95%
Chemical Name or Material Zolpidem Phenyl-4-carboxylic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.