missing translation for 'onlineSavingsMsg'
Learn More

Zolpidem Phenyl-4-carboxylic Acid (1 mg/mL in 1:1 acetonitrile:water), TRC

Product Code. 30007320
Change view
Click to view available options
Quantity:
1 x 1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007320 1 x 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30007320 Supplier Toronto Research Chemicals Supplier No. TRCKIT15251X1ML
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 109461-65-6
Molecular Formula C19 H19 N3 O3
Quantity 1 x 1 mL
Synonym Zolpidem Phenyl-4-carboxylic Acid, Benzoic acid, 4-[3-[2-(dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]-, 4-[3-[2-(Dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic acid, Imidazo[1,2-a]pyridine, benzoic acid deriv., SL 84.0589, Zolpidem carboxylic acid
InChI Formula InChI=1S/C19H19N3O3/c1-12-4-9-16-20-18(13-5-7-14(8-6-13)19(24)25)15(22(16)11-12)10-17(23)21(2)3/h4-9,11H,10H2,1-3H3,(H,24,25)
SMILES CN(C)C(=O)Cc1c(nc2ccc(C)cn12)c3ccc(cc3)C(=O)O
IUPAC Name 4-[3-[2-(dimethylamino)-2-oxoethyl]-6-methylimidazo[1,2-a]pyridin-2-yl]benzoic acid
Molecular Weight (g/mol) 337.37
Formula Weight 337.1426
Concentration or Composition (by Analyte or Components) Zolpidem Phenyl-4-carboxylic acid 1 mg/mL
Chemical Name or Material Zolpidem Phenyl-4-carboxylic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.