missing translation for 'onlineSavingsMsg'
Learn More

Zolmitriptan, TRC

Product Code. 30137428
Change view
Click to view available options
Quantity:
10 mg
50 mg
1 g
Unit Size:
10mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137428 50 mg 50mg
30003331 1 g 1g
30003928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137428 Supplier Toronto Research Chemicals Supplier No. TRCZ63900050MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 139264-17-8
Molecular Formula C16 H21 N3 O2
Molecular Weight (g/mol) 287.36
Synonym Zolmitriptan, (4S)-4-[[3-[2-Dimethylamino)ethyl]-1H-indol-5-yl]methyl]oxazolidin-2-one, (S)-4-[[3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl]methyl]-2-oxazolidinone, 311C90, Asco Top, BW 311C90, Xolnox, Zipton, Zomig, Zominat
IUPAC Name (4S)-4-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1,3-oxazolidin-2-one
SMILES CN(C)CCc1c[nH]c2ccc(C[C@H]3COC(=O)N3)cc12

Tekniske data

Quantity 50 mg
InChI Formula InChI=1S/C16H21N3O2/c1-19(2)6-5-12-9-17-15-4-3-11(8-14(12)15)7-13-10-21-16(20)18-13/h3-4,8-9,13,17H,5-7,10H2,1-2H3,(H,18,20)/t13-/m0/s1
Formula Weight 287.1634
Chemical Name or Material Zolmitriptan
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.