missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zolmitriptan
GREENER_CHOICE

Product Code. 30001535
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001535 10 mg 10mg
30001553 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001535 Supplier Tocris Bioscience™ Supplier No. 4657/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent 5-HT1B/1D/1F agonist

Potent 5-HT1B/1D/1F agonist (Ki values are 0.63, 5.01 and 63.09 nM for h5-HT1D, h5-HT1B and h5-HT1F receptors respectively). Inhibits trigeminal nerve stimulation-mediated neurogenic plasma protein extravasation in guinea pigs; blocks long excitatory postsynaptic potentials (EPSPs) in a rat model of spinal cord injury.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139264-17-8
Molecular Formula C16H21N3O2

Specifications

Quantity 10 mg
Formula Weight 287.36
Purity >99%
Molecular Weight (g/mol) 287.36
Chemical Name or Material Zolmitriptan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.