missing translation for 'onlineSavingsMsg'
Learn More

Zoledronic Acid Monohydrate, Mikromol™

Product Code. 17943576
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943576 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17943576 Supplier Mikromol Supplier No. MM1453.000250
 Restricted Item
Estimated Shipment: 20-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Zoledronic Acid Monohydrate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zoledronic Acid Monohydrate
CAS 165800-06-6
Analyte or Component Names Zoledronic Acid Monohydrate
Shipping Condition Room Temperature
Molecular Formula C5 H10 N2 O7 P2 . H2 O
Certifications/Compliance ISO 17034
Synonym Zoledronic Acid Monohydrate, P,P'-[1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid hydrate (1:1), [1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid monohydrate, Zoledronic acid hydrate, Zoledronic acid monohydrate
InChI Formula InChI=1S/C5H10N2O7P2.H2O/c8-5(15(9,10)11,16(12,13)14)3-7-2-1-6-4-7;/h1-2,4,8H,3H2,(H2,9,10,11)(H2,12,13,14);1H2
SMILES O.OC(Cn1ccnc1)(P(=O)(O)O)P(=O)(O)O
IUPAC Name (1-hydroxy-2-imidazol-1-yl-1-phosphonoethyl)phosphonic acid;hydrate
Molecular Weight (g/mol) 290.1
Quantity 250 mg
Formula Weight 290.0069 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.