missing translation for 'onlineSavingsMsg'
Learn More

Zoledronic acid monohydrate, MedChemExpress

Product Code. 16412007
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16412007

Brand: Medchem Express HY13777A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zoledronic acid monohydrate (Zoledronate monohydrate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic acid monohydrate inhibits the differentiation and apoptosis of osteoclasts. Zoledronic acid monohydrate also has anti-cancer effects.

Zoledronic acid monohydrate (Zoledronate monohydrate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic acid monohydrate inhibits the differentiation and apoptosis of osteoclasts. Zoledronic acid monohydrate also has anti-cancer effects.

Specifications

Chemical Name or Material Zoledronic acid monohydrate
CAS 165800-06-6
Color White
Purity Grade Notes Research
Molecular Formula C5H12N2O8P2
Quantity 50 mg
Synonym Zoledronate monohydrate CGP 42446 monohydrate CGP42446A monohydrate ZOL 446 monohydrate
Solubility Information H2O : 14.29 mg/mL (49.26 mM; ultrasonic and adjust pH to 8 with NaOH) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CN1C=CN=C1.[H]O[H]
Molecular Weight (g/mol) 290.1
Formula Weight 290.1
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.