missing translation for 'onlineSavingsMsg'
Learn More

Zoledronic acid monohydrate, MedChemExpress

Product Code. 16446326
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446326 100 mg 100mg
16412007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16446326 Supplier Medchem Express Supplier No. HY13777A
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Zoledronic acid monohydrate (Zoledronate monohydrate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic acid monohydrate inhibits the differentiation and apoptosis of osteoclasts. Zoledronic acid monohydrate also has anti-cancer effects.

Zoledronic acid monohydrate (Zoledronate monohydrate) is a third-generation bisphosphonate (BP), with potent anti-resorptive activity. Zoledronic acid monohydrate inhibits the differentiation and apoptosis of osteoclasts. Zoledronic acid monohydrate also has anti-cancer effects.

Specifications

Chemical Name or Material Zoledronic acid monohydrate
CAS 165800-06-6
Color White
Purity Grade Notes Research
Molecular Formula C5H12N2O8P2
Quantity 100 mg
Synonym Zoledronate monohydrate CGP 42446 monohydrate CGP42446A monohydrate ZOL 446 monohydrate
Solubility Information H2O : 14.29 mg/mL (49.26 mM; ultrasonic and adjust pH to 8 with NaOH) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble)
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CN1C=CN=C1.[H]O[H]
Molecular Weight (g/mol) 290.1
Formula Weight 290.1
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.