missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Zoledronic acid monohydrate

Product Code. 16657062 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16657062 1 g 1g
16667062 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16657062 Supplier Thermo Scientific Chemicals Supplier No. 466210010
  • ONLINE EXCLUSIVE PRICE
    £37.40
    £33.65 / 1g
    Save £3.75
    10% Off
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 165800-06-6 | C5H12N2O8P2, C5 H10 N2 O7 P2 . H2 O | 290.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165800-06-6
Molecular Formula C5 H10 N2 O7 P2 . H2 O

Specifications

Quantity 1 g
Formula Weight 290.11
Physical Form Crystalline Powder
Product Type Reagent
Purity 97.5 to 102.5% (dry substance) (Titration with NaOH)
InChI Key FUXFIVRTGHOMSO-UHFFFAOYSA-N
SMILES O.OC(CN1C=CN=C1)(P(O)(O)=O)P(O)(O)=O
IUPAC Name [1-hydroxy-2-(1H-imidazol-1-yl)-1-phosphonoethyl]phosphonic acid hydrate
Molecular Weight (g/mol) 290.11
Chemical Name or Material Zoledronic acid monohydrate
compliance-icons
Product Identifier
  • Zoledronic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Zoledronic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.