missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Zoledronic Acid (Hydrate)

Product Code. 17180242
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17180242 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17180242 Supplier STEMCELL Technologies Supplier No. 73572
 Restricted Item
Estimated Shipment: 15-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Inhibitor of bone resorption; Inhibits farnesyl diphosphate (FPP) synthase

  • Zoledronic acid is a nitrogen-containing bisphosphonate that inhibits farnesyl diphosphate (FPP) synthase (IC50 = 5 nM), thereby preventing protein prenylation and osteoclast-mediated bone resorption (Dunford et al.) In addition, it has a high affinity for hydroxyapatite (Ki = 3.5 μM) which allows it to bind directly to mineralized bone (Nancollas et al.) This product is supplied as the hydrate form of the molecule
  • Inhibits proliferation, angiogenesis, and adhesion to bone, in several cancer cell lines (Li & Davis; Zekri et al.)
  • Inhibits breast and prostate carcinoma cell invasion in vitro (Boissier et al.; Li & Davis)
  • Induces apoptosis of osteoclastoma cells in vitro (Benford et al.)

Specifications

CAS 165800-06-6
Synonym Zoledronate
Quantity 50 mg
Molecular Formula C5H10N2O7P2 · H2O
Purity ≥95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.