missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zoledronic Acid
GREENER_CHOICE

Product Code. 18711368
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18711368

Brand: Tocris Bioscience™ 6111/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent farnesyl diphosphate (FPP) synthase inhibitor

Zoledronic Acid is a potent bisphophonate farnesyl diphosphate (FPP) synthase inhibitor (IC50 = 20 nM). Inhibits osteoclast-mediated bone resorption. Also inhibits Ras signaling and tumor growth, and induces apoptosis in pancreatic cancer cells. Reverses epithelial-mesenchymal transition and inhibits breast cancer cell renewal via inactivation of NF-κB.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ras GTPase Inhibitors
CAS 118072-93-8
Synonym [1-Hydroxy-2-(1H-imidazol-1-yl)ethylidene]bisphosphonic acid
Quantity 10 mg
Solubility Soluble to 50mM in 1.1eq. NaOH and to 3mM in water
Molecular Formula C5H10N2O7P2
Formula Weight 272.09
Physical Form Solid
Color Off-white
Product Type Bioactive Small Molecule
Purity >98%
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.