missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Zolantidine dimaleate
GREENER_CHOICE

Product Code. 18015252
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18015252 10 mg 10mg
30001191 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18015252 Supplier Tocris Bioscience™ Supplier No. 1070/10
  • ONLINE EXCLUSIVE PRICE
    £173.00
    £162.75 / 10mg
    Save £10.25
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent, centrally active H2 antagonist

A potent, selective and brain penetrating H2 receptor antagonist.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104076-39-3
Molecular Formula C30H39N3O9S

Specifications

Quantity 10 mg
Formula Weight 613.68
Purity >98%
Molecular Weight (g/mol) 617.71
Chemical Name or Material Zolantidine dimaleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.