missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZO-3 Recombinant Rabbit Monoclonal Antibody (K01_2K30)
Description
ZO-3 Recombinant Monoclonal Antibody for Western Blot
ZO-3 is a member of the membrane associated guanylate kinse (MAGUK) family and are considered tight junction-associated proteins. Tight junctions are complexes of proteins that create intercellular boundaries between the plasma membrane domains of epithelial and endothelial cells. ZO-3 is believed to have both structural and signaling roles, and is characteristically defined by three protein-protein interaction modules: the PDZ domain, the SH3 domain and the guanylate kinase (GuK) domain. ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue.
Specifications
Specifications
| Antigen | ZO-3 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_2K30 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4 |
| Gene | TJP3 |
| Gene Accession No. | O95049 |
| Gene Alias | tight junction protein 3; tight junction protein 3 (zona occludens 3); tight junction protein ZO-3; TJP3; TJP-3; ZO3; ZO-3; Zona occludens protein 3; zonula occludens protein 3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?