missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZO-3 Monoclonal Antibody (K24021_24G11), Invitrogen™

Product Code. 30206125
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30206125 200 μL 200µg
30206991 50 μL 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30206125 Supplier Invitrogen™ Supplier No. MA556309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: DSDEDDGPQR VEEVDQGRGY DGDSSSGSGR SWDERSRRPR PGRRGRAGSH GRRSPGGGSE ANGLALVSGF KRLPRQDVQM KPVKSVLVKR RDSEEFGVKL GSQIFIKHIT DSGLAARHRG LQEGDLILQI NGVSSQNLSL NDTRRLIEKS EGKLSLLVLR DRGQFLVNIP PAVSDSDSSP LEDISDLASE LSQAPPSHIP PPPRHAQRSP EASQTDSPVE SPRLRRESSV DSRTISEPDE QRSELPRESS YDIYRVPSSQ SMEDRGYSPD TRVVRFLKGK SIGLRLAGGN DVGIFVSGVQ AGSPADGQGI QEGDQILQVN DVPFQNLTRE EAVQFLLGLP PGEEMELVTQ

ZO-3 is a member of the membrane associated guanylate kinse (MAGUK) family and are considered tight junction-associated proteins. Tight junctions are complexes of proteins that create intercellular boundaries between the plasma membrane domains of epithelial and endothelial cells. ZO-3 is believed to have both structural and signaling roles, and is characteristically defined by three protein-protein interaction modules: the PDZ domain, the SH3 domain and the guanylate kinase (GuK) domain. ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone K24021_24G11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TJP3
Gene Accession No. O95049
Gene Alias tight junction protein 3; tight junction protein 3 (zona occludens 3); tight junction protein ZO-3; TJP3; TJP-3; ZO3; ZO-3; Zona occludens protein 3; zonula occludens protein 3
Gene Symbols TJP3
Host Species Mouse
Immunogen Recombinant human TJP3 (immunogen range is 111-460).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.