missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZO-2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17915801
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17915801 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17915801 Supplier Invitrogen™ Supplier No. PA5143806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Guinea Pig Polyclonal Antibody

The antibody cross reacts with mouse ZO-2.

CD45 (LCA, leukocyte common antigen) is a receptor-type protein tyrosine phosphatase (PTP) ubiquitously expressed in all nucleated hematopoietic cells, comprising approximately 10% of all surface proteins in lymphocytes. CD45 is absent on non-hematopoietic cell lines, normal and malignant, non-hematopoietic tissues. CD45 glycoprotein is crucial in lymphocyte development and antigen signaling, serving as an important regulator of Src-family kinases. CD45 protein exists as multiple isoforms as a result of alternative splicing, differ in their extracellular domains but share identical transmembrane and cytoplasmic domains. CD45RA is an isoform of the CD45 complex and has restricted expression between different subtypes of lymphoid cells. CD45 isoforms differ in their ability to translocate into the glycosphingolipid-enriched membrane domains and their expression depends on cell type and physiological state of the cell. CD45 has been shown to be an essential regulator of T- and B-cell antigen receptor signaling and suppresses JAK kinases to regulate cytokine receptor signaling. CD45 is also important in promoting cell survival by modulating integrin-mediated signal transduction pathway, DNA fragmentation during apoptosis and inhibition or upregulation of various immunological functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZO-2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene TJP2
Gene Accession No. Q9UDY2
Gene Alias C9DUPq21.11; DFNA51; DUP9q21.11; Friedreich ataxia region gene X104 (tight junction protein ZO-2); PFIC4; tight junction protein; tight junction protein 2; tight junction protein 2 (zona occludens 2); tight junction protein ZO-2; Tight junction protein ZO-2 (Zonula occludens 2 protein) (Zona occludens 2 protein) (Tight junction protein 2); Tjp2; X104; ZO2; ZO-2; zona occludens 2; zona occludens protein 2; Zonula occludens protein 2
Gene Symbols TJP2
Host Species Guinea Pig
Immunogen Amino acids 411-467 of human ZO-2.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9414
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype Ig
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.