missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZO-1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA521958

Product Code. 13230169

  • £439.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Beskrivning

Beskrivning

Recommended positive controls: HCT116, 293T, A431, HeLa. Predicted reactivity: Mouse (99%), Rat (99%), Dog (84%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
TRUSTED_SUSTAINABILITY
Specifikationer

Specifikationer

ZO-1
Polyclonal
Unconjugated
TJP1
DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1
Rabbit
Antigen affinity chromatography
RUO
7082
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q07157
TJP1
Recombinant protein encompassing a sequence within the C-terminus region of human ZO-1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Produktförslag

Produktförslag

Filmer
SDS
Dokument

Dokument

Produktspecifikationer
Kampanjer

Kampanjer

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.