missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZO-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528858
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, A549, H1299, HCT116. Predicted reactivity: Mouse (100%), Rat (99%), Dog (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
Specifications
| ZO-1 | |
| Polyclonal | |
| Unconjugated | |
| TJP1 | |
| DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100736682, 21872, 407102, 7082 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.19 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P39447, Q07157 | |
| TJP1 | |
| Recombinant protein encompassing a sequence within the center region of human ZO-1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Bovine, Human, Mouse, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction