Learn More
Invitrogen™ ZO-1 Monoclonal Antibody (ZO1-1A12), Alexa Fluor™ 488
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA339100A488
Product Code. 15628028
  • £392.00 / 50µL
Description
ZO-1 Monoclonal Antibody for ICC/IF
ZO-1 forms complexes with either ZO-2 or ZO-3. In addition, these proteins can also associate with claudin, occludin and F-actin, at tight junction stands, where they provide a linkage between the actin cytoskeleton and the tight junction. ZO-1 expression is significantly reduced in many breast cancer lines. ZO-2 and ZO-3 are ubiquitously expressed within epithelial tight junctions, and unlike ZO-1, which is also expressed at cell junctions of cardiac myocytes, ZO-2 is not expressed in nonepithelial tissue. ZO-1 is a protein located on a cytoplasmic membrane surface of intercellular tight junctions. ZO-1 may be involved in signal transduction at cell-cell junctions. Two transcript variants encoding distinct isoforms have been identified for the ZO-1 gene. Diseases associated with ZO-1 dysfunction include Celiac Disease and Congenital Nephrotic Syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZO-1
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
O97758, Q07157
TJP1
Human recombinant ZO-1 fusion protein encompassing amino acids 334-634.
50 μL
Primary
Canine, Human
Antibody
IgG1
Immunocytochemistry
ZO1-1A12
Alexa Fluor 488
TJP1
DKFZp686M05161; I79_005485; MGC133289; OTTHUMP00000174520; Tight junction protein 1; tight junction protein 1 (zona occludens 1); tight junction protein ZO-1; Tjp1; zo1; ZO-1; ZO-1 MDCK; zona occludens 1; zona occludens protein 1; zonula occludens 1; zonula occludens 1 protein; zonula occludens protein 1
Mouse
Affinity chromatography
RUO
403752, 7082
4°C, store in dark, DO NOT FREEZE!