missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZnT6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13526-1-AP-150UL
This item is not returnable.
View return policy
Description
Zinc functions as a cofactor for numerous enzymes, nuclear factors, and hormones and as an intra- and intercellular signal ion. Members of the zinc transporter (ZNT)/SLC30 subfamily of the cation diffusion facilitator family, such as SLC30A6, permit cellular efflux of zinc.Specifications
| ZnT6 | |
| Polyclonal | |
| Unconjugated | |
| SLC30A6 | |
| MST103, MSTP103, SLC30A6, Zinc transporter 6, ZnT 6, ZNT6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55676 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6NXT4 | |
| SLC30A6 | |
| ZnT6 Fusion Protein Ag4403 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction