missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZnT-8/SLC30A8 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82056V
This item is not returnable.
View return policy
Description
ZnT-8/SLC30A8 Polyclonal antibody specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ZnT-8/SLC30A8 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZNT8ZnT-8 | |
| SLC30A8 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human SLC30A8. Amino Acid Squence: CHSGSKPTEKGANEY | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 169026 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction