missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZnT-8/SLC30A8 Antibody (815039), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7936V
This item is not returnable.
View return policy
Description
ZnT-8/SLC30A8 Monoclonal antibody specifically detects ZnT-8/SLC30A8 in Human samples. It is validated for Immunohistochemistry, Flow Cytometry, CyTOF
Specifications
| ZnT-8/SLC30A8 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Flow Cytometry, CyTOF | |
| 815039 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| solute carrier family 30 (zinc transporter), member 8, Solute carrier family 30 member 8, zinc transporter 8, zinc transporter ZnT-8, ZNT8ZnT-8 | |
| E. coli-derived recombinant human ZnT-8, Lys268-Pro359, Accession # Q8IWU4 | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 169026 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction