Learn More
Specac™ ZnSe Demountable Heatable Liquid Flow Cell VTCH Compatible
Description
This cell is designed for use in the Specac variable temperature cell GS21525. This is a demountable heatable liquid flow cell with ZnSe windows and choice of pathlength from 0.025mm up to 1.0mm. This cell is fitted with 1/16in. Swagelok fittings for flow purposes.
Specifications
Specifications
| Type | ZnSe Demountable Heatable Liquid Flow Cell |
| Description | Pathlength from 0.025 mm up to 1.0 mm |