missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNRF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £395.00
Specifications
| Antigen | ZNRF3 |
|---|---|
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18356264
|
Novus Biologicals
NBP3-03924-20UL |
20 μL |
£178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18022926
|
Novus Biologicals
NBP3-03924-100UL |
100 μL |
£395.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNRF3 Polyclonal specifically detects ZNRF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ZNRF3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 84133 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| novel C3HC4 type Zinc finger (ring finger), zinc and ring finger 3, zinc/RING finger protein 3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human ZNRF3 (NP_115549.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title