missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ZNRF2 Antibody (SR3261), Novus Biologicals™
Rabbit Monoclonal Antibody
£240.00 - £290.00
Specifications
| Antigen | ZNRF2 |
|---|---|
| Clone | SR3261 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041770
|
Novus Biologicals™
NBP3-27761-100ul |
100 μL |
£290.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041704
|
Novus Biologicals™
NBP3-27761-50ul |
50 μL |
£240.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNRF2 Monoclonal antibody specifically detects ZNRF2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western BlotSpecifications
| ZNRF2 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 6.3.2.-, Protein Ells2, RING finger protein 202, RNF202E3 ubiquitin-protein ligase ZNRF2, zinc and ring finger 2, zinc finger/RING finger 2, Zinc/RING finger protein 2 | |
| A synthesized peptide derived from human ZNRF2 (Uniprot #: Q8NHG8) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| SR3261 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Zinc Finger | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 223082 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title