missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNRF2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28715
This item is not returnable.
View return policy
Description
ZNRF2 Polyclonal specifically detects ZNRF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ZNRF2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| EC 6.3.2.-, Protein Ells2, RING finger protein 202, RNF202E3 ubiquitin-protein ligase ZNRF2, zinc and ring finger 2, zinc finger/RING finger 2, Zinc/RING finger protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:1000-1:5000, Immunoprecipitation 10 ug/mg lysate | |
| NP_667339.1 | |
| ZNRF2 | |
| The immunogen recognized by this antibody maps to a region between residue 192 and 242 of human zinc and ring finger 2 using the numbering given in entry NP_667339.1 (GeneID 223082). | |
| 100 μL | |
| Zinc Finger | |
| 223082 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction