missing translation for 'onlineSavingsMsg'
Learn More

ZNRF2 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18432168 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18432168 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18432168 Supplier Novus Biologicals Supplier No. NBP128697AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNRF2 Polyclonal antibody specifically detects ZNRF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNRF2
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.-, Protein Ells2, RING finger protein 202, RNF202E3 ubiquitin-protein ligase ZNRF2, zinc and ring finger 2, zinc finger/RING finger 2, Zinc/RING finger protein 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 100 and 150 of human zinc and ring finger 2 using the numbering given in entry NP_667339.1 (GeneID 223082).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 223082
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.