missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNRF1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA537982
This item is not returnable.
View return policy
Description
This antibody is predicted to react with human, rat, dog, pig and cow based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
E3 ubiquitin-protein ligase that mediates the ubiquitination of AKT1 and GLUL, thereby playing a role in neuron cells differentiation. Plays a role in the establishment and maintenance of neuronal transmission and plasticity. Regulates Schwann cells differentiation by mediating ubiquitination of GLUL. Promotes neurodegeneration by mediating 'Lys-48'-linked polyubiquitination and subsequent degradation of AKT1 in axons: degradation of AKT1 prevents AKT1-mediated phosphorylation of GSK3B, leading to GSK3B activation and phosphorylation of DPYSL2/CRMP2 followed by destabilization of microtubule assembly in axons (Probable).
Specifications
| ZNRF1 | |
| Polyclonal | |
| Unconjugated | |
| ZNRF1 | |
| B830022L21Rik; E3 ubiquitin-protein ligase ZNRF1; MGC101991; nerve injury gene 283; nerve injury-induced gene 283 protein; Nin283; peripheral nerve injury protein nin283; ring finger protein 42; RING-type E3 ubiquitin transferase ZNRF1; Rnf42; zgc:77896; zinc and ring finger 1; zinc and ring finger 1, E3 ubiquitin protein ligase; zinc and ring finger protein 1; zinc ring finger protein 1; zinc/RING finger protein 1; ZNRF1; Zrfp1 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 170737, 84937 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q8ND25, Q91V17 | |
| ZNRF1 | |
| Peptide with sequence C-SDSTYAHGNGYQET, from the internal region of mouse Znrf1 (aa 93-107). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction