missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNHIT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553736
This item is not returnable.
View return policy
Description
Immunogen sequence: KEELMHGECV KEEKDFLKKE IVDDTKVKEE PPINHPVGCK RKLAMSRCET CGTEEAKYRC PRCMRYSCSL PCVKKHKAEL TCNGVRDKTA YISIQQFTEM NLL Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 72%.
Required for box C/D snoRNAs accumulation involved in snoRNA processing, snoRNA transport to the nucleolus and ribosome biogenesis. [UniProt].
Specifications
| ZNHIT6 | |
| Polyclonal | |
| Unconjugated | |
| ZNHIT6 | |
| 2410019A14Rik; BCD1; box C/D snoRNA essential 1 homolog; box C/D snoRNA protein 1; box C/d snoRNA protein 1-like protein; C1orf181; C3H1ORF181; FLJ20729; NY-BR-75; RGD1308723; Serologically defined breast cancer antigen NY-BR-75; zinc finger HIT domain-containing protein 6; zinc finger HIT-type containing 6; zinc finger, HIT type 6; zinc finger, HIT-type containing 6; Znhit6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 54680 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NWK9 | |
| ZNHIT6 | |
| Recombinant protein corresponding to Human ZNHIT6. Recombinant protein control fragment (Product #RP-91618). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction