Learn More
ZNFX1 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55381-1-AP-150UL
This item is not returnable.
View return policy
Description
The ZNFX1 antibody from Proteintech is a rabbit polyclonal antibody generated with peptide of human ZNFX1. This antibody recognizes human antigen. The ZNFX1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZNFX1 | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| KIAA1404, ZNFX1 | |
| 150 μL | |
| 57169 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |