missing translation for 'onlineSavingsMsg'
Learn More

ZNF96 Antibody (3E4), Novus Biologicals™

Product Code. 18399948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18399948 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18399948 Supplier Novus Biologicals Supplier No. H00009753M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZNF96 Monoclonal antibody specifically detects ZNF96 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF96
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3E4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH41661
Gene Alias KIAA0426dJ29K1.2, ZFP96, zinc finger and SCAN domain containing 12, Zinc finger protein 305ZNF305, Zinc finger protein 96ZNF96zinc finger and SCAN domain-containing protein 12, ZNF29K1
Host Species Mouse
Immunogen ZNF96 (AAH41661, 1 a.a. ~ 604 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASTWAIQAHMDQDEPLEVKIEEEKYTTRQDWDLRKNNTHSREVFRQYFRQFCYQETSGPREALSRLRELCHQWLRPETHTKEQILELLVLEQFLTILPEELQAWVQEQHPESGEEVVTVLEDLERELDEPGEQVSVHTGEQEMFLQETVRLRKEGEPSMSLQSMKAQPKYESPELESQQEQVLDVETGNEYGNLKQEVSEEMEPHGKTSSKFENDMSKSARCGETREPEEITEEPSACSREDKQPTCDENGVSLTENSDHTEHQRICPGEESYGCDDCGKAFSQHSHLIEHQRIHTGDRPYKCEECGKAFRGRTVLIRHKIIHTGEKPYKCNECGKAFGRWSALNQHQRLHTGEKHYHCNDCGKAFSQKAGLFHHIKIHTRDKPYQCTQCNKSFSRRSILTQHQGVHTGAKPYECNECGKAFVYNSSLVSHQEIHHKEKCYQCKECGKSFSQSGLIQHQRIHTGEKPYKCDVCEKAFIQRTSLTEHQRIHTGERPYKCDKCGKAFTQRSVLTEHQRIHTGERPYKCDECGNAFRGITSLIQHQRIHTGEKPYQCDECGKAFRQRKKTSYKEILLKNHSEPQAGVNLLLSSLIPEWQSCFRRDL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9753
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.