missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF91 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543064
This item is not returnable.
View return policy
Description
Peptide sequence: HKIIHAGEKL YKCEECGKAF NRSSNLTIHK FIHTGEKPNW Sequence homology: Human: 100%; Mouse: 93%; Rat: 93%.
ZNF91 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 36 C2H2-type zinc fingers and 1 KRAB domain. The function of the ZNF91 protein remains unknown. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Specifications
| ZNF91 | |
| Polyclonal | |
| Unconjugated | |
| ZNF91 | |
| HPF7; HTF10; Zinc finger protein 91; zinc finger protein 91 (HPF7, HTF10); zinc finger protein HPF7; Zinc finger protein HTF10; ZNF91 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7644 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q05481 | |
| ZNF91 | |
| Synthetic peptide directed towards the N-terminal of human ZNF91. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction