missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF891 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ZNF891 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF891 Polyclonal specifically detects ZNF891 in Human samples. It is validated for Western Blot.Specifications
| ZNF891 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 440122 | |
| Synthetic peptide directed towards the middle region of human hCG_1646157. Peptide sequence PVPNNLHMAQNACECNKDETLCHQSSLKKQGQTHTEKKHECNQCGKAFKR. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ZNF891 zinc finger protein 891 | |
| ZNF891 | |
| IgG | |
| 63 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title