missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF879 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543718
This item is not returnable.
View return policy
Description
Peptide sequence: MKSGGTNAGG SARETRRLSG AQAQLRPAAT GNVSELGVPL ASCAVWSCAP.
The exact functions of DKFZp686E2433 remain unknown.
Specifications
| ZNF879 | |
| Polyclonal | |
| Unconjugated | |
| ZNF879 | |
| DKFZp686E2433; Zinc finger protein 879; ZNF879 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 345462 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| B4DU55 | |
| ZNF879 | |
| Synthetic peptide directed towards the N-terminal of human DKFZp686E2433. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction