missing translation for 'onlineSavingsMsg'
Learn More

ZNF879 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£381.00

Specifications

Antigen ZNF879
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18321640
View Documents
Novus Biologicals
NBP1-91425
100 μL
£381.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF879 Polyclonal specifically detects ZNF879 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF879
Polyclonal
Rabbit
Human
345462
Synthetic peptide directed towards the N terminal of human DKFZp686E2433. Peptide sequence: ATGNVSELGVPLASCAVWSCAPPAEAGPGAFSKRGSREGEMARRLLPAHV
Primary
Western Blot
Unconjugated
RUO
zinc finger protein 879
ZNF879
IgG
77 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.