missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF846 Antibody (ZNF846/2687), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08846AF350
This item is not returnable.
View return policy
Description
ZNF846 Monoclonal antibody specifically detects ZNF846 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array
Specifications
| ZNF846 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| zinc finger protein 846 | |
| Recombinant full-length human ZNF846 protein (Uniprot: Q147U1) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin), Protein Array | |
| ZNF846/2687 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 162993 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction