missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF846 Antibody (PCRP-ZNF846-1E12) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20736JF585
This item is not returnable.
View return policy
Description
ZNF846 Monoclonal antibody specifically detects ZNF846 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF846 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| zinc finger protein 846 | |
| Recombinant full-length human ZNF846 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| PCRP-ZNF846-1E12 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 162993 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction