missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF81 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551625
This item is not returnable.
View return policy
Description
Immunogen sequence: VHPSPNLILS QKRPHKRDSF GKSFKHNLDL HIHNKSNAAK NLDKTIGHGQ VFTQNSSYSH HENTHTGVKF CERNQCGKVL SLKHSLSQNV KFPIGEKANT CT Highest antigen sequence identity to the following orthologs: Mouse - 34%, Rat - 50%.
This gene encodes a protein that likely functions as a transcription factor. The protein contains an N-terminal KRAB domain and several C2H2-type zinc finger motifs. Mutations in this gene cause mental retardation X-linked type 45 (MRX45).
Specifications
| ZNF81 | |
| Polyclonal | |
| Unconjugated | |
| ZNF81 | |
| dJ54B20.6; HFZ20; MRX45; zinc finger protein 41-like; zinc finger protein 81; ZNF81 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 347344 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51508 | |
| ZNF81 | |
| Recombinant protein corresponding to Human ZNF81. Recombinant protein control fragment (Product #RP-91998). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction