missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF787 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78197
This item is not returnable.
View return policy
Description
ZNF787 Polyclonal specifically detects ZNF787 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ZNF787 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| TIP20, transcription termination factor I interacting peptide 20, TTF-I interacting peptide 20, TTF-I-interacting peptide 20, zinc finger protein 787 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 126208 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 mcg/mg | |
| NP_001002836.2 | |
| ZNF787 | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Zinc Finger Protein 787 using the numbering given in entry NP_001002836.2 (GeneID 126208). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction