missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF786 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA554435

Product Code. 15712371

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: EGIPGPRNLD LPGLWDVPAW ESTQHPWPVC GESCWENNHL VMHQRGHSKD RTRRAWEKFN KRAETQMPWS SPRVQRH Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 40%.

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krüppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF786 (zinc finger protein 786) is a 782 amino acid protein that belongs to the Krüppel C2H2-type zinc-finger protein family and is thought to function in transcriptional regulation. Localizing to nucleus, ZNF786 contains 16 C2H2-type zinc fingers, a single KRAB domain and is encoded by a gene that maps to human chromosome 7q36.1.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF786
Polyclonal
Unconjugated
ZNF786
A730012O14Rik; Zfp786; Zinc finger protein 786; Znf786
Rabbit
Antigen affinity chromatography
RUO
136051
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8N393
ZNF786
Recombinant protein corresponding to Human ZNF786. Recombinant protein control fragment (Product #RP-92846).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.