missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF784 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569741
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 85%; Rat: 100%; Zebrafish: 91%.
ZNF784 may be involved in transcriptional regulation.
Specifications
| ZNF784 | |
| Polyclonal | |
| Unconjugated | |
| ZNF784 | |
| C730049F21Rik; Zfp784; Zinc finger protein 784; Znf784 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 147808 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NCA9 | |
| ZNF784 | |
| synthetic peptide directed towards the middle region of human ZNF784. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction