missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF782 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543545
This item is not returnable.
View return policy
Description
Peptide sequence: TNKLLTTEQE ISGKPHNRDI NIFRARMMPC KCDIAGSACQ GLSLMAPHCQ Sequence homology: Dog: 79%; Human: 100%.
ZNF782 may be involved in transcriptional regulation.
Specifications
| ZNF782 | |
| Polyclonal | |
| Unconjugated | |
| ZNF782 | |
| Zinc finger protein 782; ZNF782 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 158431 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6ZMW2 | |
| ZNF782 | |
| Synthetic peptide directed towards the N-terminal of human ZNF782 (aa 107-156). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction