missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF768 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78220
This item is not returnable.
View return policy
Description
ZNF768 Polyclonal specifically detects ZNF768 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZNF768 | |
| Polyclonal | |
| Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| NP_078947.3 | |
| ZNF768 | |
| The immunogen for this product maps to a region between residue 1 and 15 of human zinc finger protein 768 using the numbering given in entry NP_078947.3 (GeneID 79724). | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| zinc finger protein 768 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 79724 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction