missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF766 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£169.00 - £255.00
Specifications
| Antigen | ZNF766 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18664552
|
Novus Biologicals
NBP2-98465-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650325
|
Novus Biologicals
NBP2-98465-50ul |
50 μg |
£169.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF766 Polyclonal antibody specifically detects ZNF766 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| ZNF766 | |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| zinc finger protein 766 | |
| Produced in rabbits immunized with E. coli-derived Human ZNF766 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate, Immunohistochemistry-Paraffin 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.0) | |
| 90321 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title