missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF750 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21752-1-AP-150UL
This item is not returnable.
View return policy
Description
Zinc finger protein 750 is a new candidate transcription factor.Specifications
| ZNF750 | |
| Polyclonal | |
| Unconjugated | |
| ZNF750 | |
| Protein ZNF750, Zfp750, zinc finger protein 750, ZNF750 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319530, 79755 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.44 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q32MQ0, Q8BH05 | |
| Znf750 | |
| ZNF750 Fusion Protein Ag16116 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction