missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF746 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67463-1-IG-150UL
This item is not returnable.
View return policy
Description
The ZNF746 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZNF746. This antibody recognizes human antigen. The ZNF746 antibody has been validated for the following applications: WB, ELISA analysis.Specifications
| ZNF746 | |
| Monoclonal | |
| 1100 μg/mL | |
| FLJ31413 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 85-90 kDa | |
| Liquid |
| Western Blot, ELISA | |
| 3A12C5 | |
| Unconjugated | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 155061 | |
| Store at -20°C. | |
| Monoclonal Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction