missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF740 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25411-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| ZNF740 | |
| Polyclonal | |
| Unconjugated | |
| ZNF740 | |
| TB7, Zfp740, zinc finger protein 740, ZNF740 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 283337, 68744 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6NZQ6, Q8NDX6 | |
| ZNF740 | |
| ZNF740 Fusion Protein Ag22295 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction