missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF736 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA546696
This item is not returnable.
View return policy
Description
Peptide sequence: HDIKDSFQKV ILRKYGSCDL NNLHLKKDYQ SVGNCKGQKS SYNGLHQCLS Sequence homology: Human: 100%.
ZNF736 may be involved in transcriptional regulation.
Specifications
| ZNF736 | |
| Polyclonal | |
| Unconjugated | |
| ZNF736 | |
| KRAB-containing zinc-finger repressor protein; zinc finger protein 736; ZNF736 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 728927 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| B4DX44 | |
| ZNF736 | |
| Synthetic peptide directed towards the N-terminal of human ZNF736 (aa 89-138). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction