missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF732 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568705
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Human: 100%; Mouse: 77%.
This gene encodes a kruppel-associated box-containing zinc finger protein (KRAB-ZFP). The encoded protein contains an N-terminal kruppel-associated box (KRAB) domain and sixteen C-terminal C2H2-type zinc finger domains. The KRAB-ZFPs represent the largest family of mammalian transcriptional repressors, which function through the recruitment of the nuclear co-factor KRAB-Associated Protein 1 (KAP1), to engage histone modifiers and induce heterochromatin formation.
Specifications
| ZNF732 | |
| Polyclonal | |
| Unconjugated | |
| ZNF732 | |
| Zinc finger protein 732; zinc finger protein LOC654254; ZNF732 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 654254 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| B4DXR9 | |
| ZNF732 | |
| synthetic peptide directed towards the N-terminal region of human ZNF732. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction