missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZNF711 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595865
This item is not returnable.
View return policy
Description
Immunogen sequence: SEDYLMISLD DVGEKLEHMG NTPLKIGSDG SQEDAKEDGF GSEVIKVYIF KAEAEDDVEI GGTEIVTESE YTSGHSVAGV LDQSRMQREK MVYMAVKDSS Q.
This gene encodes a zinc finger protein of unknown function. It bears similarity to a zinc finger protein which acts as a transcriptional activator. This gene lies in a region of the X chromosome which has been associated with mental retardation.
Specifications
| ZNF711 | |
| Polyclonal | |
| Unconjugated | |
| ZNF711 | |
| 2810409C01Rik; A230078I01Rik; CMPX1; dJ75N13.1; dJ75N13.1 (znf6-like); MRX97; Zfp711; Zinc finger protein 6; zinc finger protein 6 (CMPX1); zinc finger protein 711; ZNF4; ZNF5; ZNF6; ZNF711 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7552 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9Y462 | |
| ZNF711 | |
| A synthetic peptide corresponding to a sequence within amino acids 200-300 of human ZNF711 (NP_0013175031). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction