missing translation for 'onlineSavingsMsg'
Learn More

ZNF703 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18408516 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408516 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408516 Supplier Novus Biologicals Supplier No. NBP232138JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF703 Polyclonal antibody specifically detects ZNF703 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF703
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 100 to 150 of human Zinc Finger Protein 703 using the numbering given in entry NP_079345.1 (GeneID 80139).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80139
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.